Product Details

SNP ID
rs200414299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11948481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCAGAGGTGACACTGGACACAAG[A/G]CATATGAGTATCAGGAATATGGACC
Phenotype
MIM: 194543
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF69 PubMed Links

Gene Details

Gene
ZNF69
Gene Name
zinc finger protein 69
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321163.1 609 Intron NP_001308092.1
NM_021915.3 609 Intron NP_068734.1
XM_006722873.3 609 Intron XP_006722936.1
XM_006722874.3 609 Intron XP_006722937.1
XM_017027231.1 609 Intron XP_016882720.1
Gene
ZNF700
Gene Name
zinc finger protein 700
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271848.1 609 Missense Mutation ACA,GCA T156A NP_001258777.1
NM_144566.2 609 Missense Mutation ACA,GCA T153A NP_653167.1

View Full Product Details