Product Details

SNP ID
rs201042020
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:52633542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGAGCGCAGAGCAGGAGGTGGTG[C/G]GCGGCTTCAGCTGCCCGCGGCCGGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM159A PubMed Links

Gene Details

Gene
FAM159A
Gene Name
family with sequence similarity 159 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042693.2 199 Missense Mutation CGC,GGC R17G NP_001036158.1
XM_011541377.2 199 Intron XP_011539679.1
XM_011541378.2 199 Intron XP_011539680.1

View Full Product Details