Product Details

SNP ID
rs200099360
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:58782420 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGGCCTGCAGCCGCGGGTGCTG[C/G]ACGGGCATCTGCTGGGGCAGGTGGT
Phenotype
MIM: 165160
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
JUN PubMed Links

Gene Details

Gene
JUN
Gene Name
Jun proto-oncogene, AP-1 transcription factor subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002228.3 1694 Silent Mutation GTC,GTG V217V NP_002219.1
Gene
LINC01135
Gene Name
long intergenic non-protein coding RNA 1135
There are no transcripts associated with this gene.

View Full Product Details