Product Details

SNP ID
rs200967848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27372689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAAAGAAGCTTTTCTCTCTGACT[C/T]CTGCTGAGGTAGAACAGAGGAGAGA
Phenotype
MIM: 606687 MIM: 605119 MIM: 605963 MIM: 613598
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF2B4 PubMed Links

Gene Details

Gene
EIF2B4
Gene Name
eukaryotic translation initiation factor 2B subunit delta
There are no transcripts associated with this gene.

Gene
PPM1G
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1G
There are no transcripts associated with this gene.

Gene
SNX17
Gene Name
sorting nexin 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267059.1 417 Missense Mutation CCT,TCT P57S NP_001253988.1
NM_001267060.1 417 Missense Mutation CCT,TCT P44S NP_001253989.1
NM_001267061.1 417 Missense Mutation CCT,TCT P49S NP_001253990.1
NM_014748.3 417 Missense Mutation CCT,TCT P69S NP_055563.1
XM_011533203.1 417 Intron XP_011531505.1
XM_017005405.1 417 Intron XP_016860894.1
Gene
ZNF513
Gene Name
zinc finger protein 513
There are no transcripts associated with this gene.

View Full Product Details