Product Details

SNP ID
rs201545588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27373938 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGGCAGAAAGTTCTGGTCAACGT[A/G]CTAACTTCAGATCAGACTGAGGATG
Phenotype
MIM: 606687 MIM: 605119 MIM: 605963 MIM: 613598
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF2B4 PubMed Links

Gene Details

Gene
EIF2B4
Gene Name
eukaryotic translation initiation factor 2B subunit delta
There are no transcripts associated with this gene.

Gene
PPM1G
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1G
There are no transcripts associated with this gene.

Gene
SNX17
Gene Name
sorting nexin 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267059.1 421 Silent Mutation GTA,GTG V121V NP_001253988.1
NM_001267060.1 421 Silent Mutation GTA,GTG V108V NP_001253989.1
NM_001267061.1 421 Silent Mutation GTA,GTG V113V NP_001253990.1
NM_014748.3 421 Silent Mutation GTA,GTG V133V NP_055563.1
XM_011533203.1 421 UTR 5 XP_011531505.1
XM_017005405.1 421 UTR 5 XP_016860894.1
Gene
ZNF513
Gene Name
zinc finger protein 513
There are no transcripts associated with this gene.

View Full Product Details