Product Details
- SNP ID
-
rs201314635
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:36028183 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGTCTCAGGAGAGGCCCACGTTCT[A/G]CCGGCAGGAGCTGAACAAGACAATC
- Phenotype
-
MIM: 600289
MIM: 608480
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MAPK14
PubMed Links
Gene Details
- Gene
- MAPK14
- Gene Name
- mitogen-activated protein kinase 14
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001315.2 |
549 |
Missense Mutation |
TAC,TGC |
Y9C |
NP_001306.1 |
NM_139012.2 |
549 |
Missense Mutation |
TAC,TGC |
Y9C |
NP_620581.1 |
NM_139013.2 |
549 |
Missense Mutation |
TAC,TGC |
Y9C |
NP_620582.1 |
NM_139014.2 |
549 |
Missense Mutation |
TAC,TGC |
Y9C |
NP_620583.1 |
XM_006714998.3 |
549 |
Intron |
|
|
XP_006715061.1 |
XM_011514310.2 |
549 |
UTR 5 |
|
|
XP_011512612.2 |
XM_017010299.1 |
549 |
Missense Mutation |
TAC,TGC |
Y9C |
XP_016865788.1 |
XM_017010300.1 |
549 |
Missense Mutation |
TAC,TGC |
Y9C |
XP_016865789.1 |
XM_017010301.1 |
549 |
Missense Mutation |
TAC,TGC |
Y9C |
XP_016865790.1 |
XM_017010302.1 |
549 |
Missense Mutation |
TAC,TGC |
Y9C |
XP_016865791.1 |
XM_017010303.1 |
549 |
Missense Mutation |
TAC,TGC |
Y9C |
XP_016865792.1 |
XM_017010304.1 |
549 |
Missense Mutation |
TAC,TGC |
Y9C |
XP_016865793.1 |
- Gene
- SLC26A8
- Gene Name
- solute carrier family 26 member 8
There are no transcripts associated with this gene.
View Full Product Details