Product Details

SNP ID
rs199705089
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:108884090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCGTAGATATGCACCTTTCACTT[A/G]AGATATGTATCCCTCATGAATTCTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C7orf66 PubMed Links

Gene Details

Gene
C7orf66
Gene Name
chromosome 7 open reading frame 66
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024607.2 320 Nonsense Mutation CAA,TAA Q89* NP_001019778.1

View Full Product Details