Product Details
- SNP ID
-
rs2278671
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:84495644 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCGCGCGCGACAGGGGCGGTACCTA[G/T]CCCTCCAGGTATGCAAGGTCCACTC
- Phenotype
-
MIM: 602563
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NKX6-1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117620724] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NKX6-1
- Gene Name
- NK6 homeobox 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006168.2 |
|
Intron |
|
|
NP_006159.2 |
View Full Product Details