Product Details
- SNP ID
-
rs11585316
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
38
- Location
-
Chr.1:52637952 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GAATCTCTAACTTCACTGAAAGATA[G/T]AACTCATTCGTGTGTCCTGCACAAG
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM159A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs56246079] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM159A
- Gene Name
- family with sequence similarity 159 member A
View Full Product Details