Product Details

SNP ID
rs2321271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:140536884 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGGACATTTCAGAGATAGAAAGG[A/T]TTAGCGTATGGGGATCTTTTGATAC
Phenotype
MIM: 610196
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ELMOD2 PubMed Links

Gene Details

Gene
ELMOD2
Gene Name
ELMO domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153702.3 Intron NP_714913.1
XM_005262885.3 Intron XP_005262942.1
XM_005262886.2 Intron XP_005262943.1
XM_011531818.2 Intron XP_011530120.1
XM_011531819.2 Intron XP_011530121.1

View Full Product Details