Product Details

SNP ID
rs2276131
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66003113 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAACTGTTTTCTCCTTTCAGGAT[A/G]AGGGGGATCGAGCAAGCCACTGTAA
Phenotype
MIM: 603811 MIM: 601891
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
BANF1 PubMed Links

Gene Details

Gene
BANF1
Gene Name
barrier to autointegration factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143985.1 Intron NP_001137457.1
NM_003860.3 Intron NP_003851.1
XM_017018514.1 Intron XP_016874003.1
XM_017018515.1 Intron XP_016874004.1
Gene
CST6
Gene Name
cystatin E/M
There are no transcripts associated with this gene.

Gene
EIF1AD
Gene Name
eukaryotic translation initiation factor 1A domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242481.1 Intron NP_001229410.1
NM_001242482.1 Intron NP_001229411.1
NM_001242483.1 Intron NP_001229412.1
NM_001242484.1 Intron NP_001229413.1
NM_001242485.1 Intron NP_001229414.1
NM_001242486.1 Intron NP_001229415.1
NM_032325.3 Intron NP_115701.2
XM_017018412.1 Intron XP_016873901.1

View Full Product Details