Product Details

SNP ID
rs2483104
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:41317518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGATAGACATGAAAATAATGATC[C/T]TGTAGCATAGCAAAGCTGGTTGAGA
Phenotype
MIM: 604601
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTRF1 PubMed Links

Gene Details

Gene
MTRF1
Gene Name
mitochondrial translational release factor 1
There are no transcripts associated with this gene.

Gene
NAA16
Gene Name
N(alpha)-acetyltransferase 16, NatA auxiliary subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001110798.1 Intron NP_001104268.1
NM_018527.3 Intron NP_060997.2
NM_024561.4 Intron NP_078837.3
XM_005266523.2 Intron XP_005266580.1
XM_006719866.2 Intron XP_006719929.1
XM_011535227.2 Intron XP_011533529.1
XM_011535228.1 Intron XP_011533530.1
XM_017020744.1 Intron XP_016876233.1
XM_017020745.1 Intron XP_016876234.1
XM_017020746.1 Intron XP_016876235.1

View Full Product Details