Product Details

SNP ID
rs2157928
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:87629088 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTGCGCCAGCCAAATCTGTGAGC[A/G]CGCAGCTCCTTGGACAGGGGCCCGG
Phenotype
MIM: 171050
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCB1 PubMed Links

Gene Details

Gene
ABCB1
Gene Name
ATP binding cassette subfamily B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000927.4 Intron NP_000918.2
Gene
RUNDC3B
Gene Name
RUN domain containing 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134405.1 Intron NP_001127877.1
NM_001134406.1 Intron NP_001127878.1
NM_138290.2 Intron NP_612147.1
XM_005250156.2 Intron XP_005250213.1
XM_005250158.2 Intron XP_005250215.1
XM_011515826.1 Intron XP_011514128.1
XM_011515827.1 Intron XP_011514129.1
XM_011515828.2 Intron XP_011514130.1
XM_011515829.2 Intron XP_011514131.1
XM_017011761.1 Intron XP_016867250.1
XM_017011762.1 Intron XP_016867251.1
XM_017011763.1 Intron XP_016867252.1
XM_017011764.1 Intron XP_016867253.1
XM_017011765.1 Intron XP_016867254.1
XM_017011766.1 Intron XP_016867255.1

View Full Product Details