Product Details

SNP ID
rs2274968
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:124869501 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCCTTCTCACCTTCCCACCTTCCC[A/G]CCTTCTCGGGCCCTTTCGGGCGGGC
Phenotype
MIM: 602502
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARPC5L PubMed Links

Gene Details

Gene
ARPC5L
Gene Name
actin related protein 2/3 complex subunit 5 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030978.2 Intron NP_112240.1
XM_011519067.1 Intron XP_011517369.1
Gene
GOLGA1
Gene Name
golgin A1
There are no transcripts associated with this gene.

Gene
RPL35
Gene Name
ribosomal protein L35
There are no transcripts associated with this gene.

View Full Product Details