Product Details

SNP ID
rs2244156
Assay Type
Functionally tested
NCBI dbSNP Submissions
62
Location
Chr.1:51910005 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCATTTAATCCTTTCAACAGCCT[C/T]GTGAAGTAGATAGTATTACTTCATT
Phenotype
MIM: 179510
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB3B PubMed Links

Gene Details

Gene
RAB3B
Gene Name
RAB3B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002867.3 10795 UTR 3 NP_002858.2
XM_017001958.1 10795 Intron XP_016857447.1

View Full Product Details