Product Details

SNP ID
rs61746162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8731099 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTCTTCAGTCACTCAGGATCAC[A/G]CCAGCTCCTCTTCTCCCTGGTGGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2Z1 PubMed Links

Gene Details

Gene
OR2Z1
Gene Name
olfactory receptor family 2 subfamily Z member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004699.2 146 Missense Mutation CAC,CGC H24R NP_001004699.1

View Full Product Details