Product Details

SNP ID
rs4317843
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:247037316 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTAGGAAAATTGCATACATTCTA[A/G]TCTTCCTTACATGTGTTGGGTTTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF670 PubMed Links

Gene Details

Gene
ZNF670
Gene Name
zinc finger protein 670
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204220.1 1520 UTR 3 NP_001191149.1
NM_033213.4 1520 UTR 3 NP_149990.1
Gene
ZNF670-ZNF695
Gene Name
ZNF670-ZNF695 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details