Product Details

SNP ID
rs2292506
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52544536 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGGGCCCGGGGCAGTCTTCTCTAC[C/T]GACTGGTTTTCTTGGAGGGTGCACT
Phenotype
MIM: 608245
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT71 PubMed Links

Gene Details

Gene
KRT71
Gene Name
keratin 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033448.2 1638 Missense Mutation CAG,CGG Q523R NP_258259.1
XM_017018749.1 1638 Missense Mutation CAG,CGG Q441R XP_016874238.1

View Full Product Details