Product Details

SNP ID
rs4498095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:143885468 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAACAGCTACTGCTCACAGTAAA[A/T]GAGCACTTGGTGGAAATTTTGTTCT
Phenotype
MIM: 138590
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GYPE PubMed Links

Gene Details

Gene
GYPE
Gene Name
glycophorin E (MNS blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002102.3 Intron NP_002093.2
NM_198682.2 Intron NP_941391.2
XM_017008139.1 Intron XP_016863628.1
XM_017008140.1 Intron XP_016863629.1

View Full Product Details