Product Details

SNP ID
rs279253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11942381 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTGCCTATTAATTCATAGGACTC[C/T]GTTATGACAGGTGCTACAGACCCAC
Phenotype
MIM: 194543
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF69 PubMed Links

Gene Details

Gene
ZNF69
Gene Name
zinc finger protein 69
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321163.1 Intron NP_001308092.1
NM_021915.3 Intron NP_068734.1
XM_006722873.3 Intron XP_006722936.1
XM_006722874.3 Intron XP_006722937.1
XM_017027231.1 Intron XP_016882720.1
Gene
ZNF700
Gene Name
zinc finger protein 700
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271848.1 Intron NP_001258777.1
NM_144566.2 Intron NP_653167.1

View Full Product Details