Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001281437.1 | Intron | NP_001268366.1 | |||
NM_001281438.1 | Intron | NP_001268367.1 | |||
NM_007002.3 | Intron | NP_008933.2 | |||
NM_175573.2 | Intron | NP_783163.1 | |||
XM_005260257.1 | Intron | XP_005260314.1 | |||
XM_011528503.1 | Intron | XP_011526805.1 | |||
XM_017027602.1 | Intron | XP_016883091.1 | |||
XM_017027603.1 | Intron | XP_016883092.1 | |||
XM_017027604.1 | Intron | XP_016883093.1 | |||
XM_017027605.1 | Intron | XP_016883094.1 | |||
XM_017027606.1 | Intron | XP_016883095.1 | |||
XM_017027607.1 | Intron | XP_016883096.1 | |||
XM_017027608.1 | Intron | XP_016883097.1 | |||
XM_017027609.1 | Intron | XP_016883098.1 | |||
XM_017027610.1 | Intron | XP_016883099.1 |