Product Details
- SNP ID
-
rs12880915
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:77983084 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTATTATTCTAACAGCTTATGTGTA[G/T]CAATTCATCCAATTACCCTATGAGG
- Phenotype
-
MIM: 600567
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NRXN3
PubMed Links
Gene Details
- Gene
- NRXN3
- Gene Name
- neurexin 3
View Full Product Details