Product Details

SNP ID
rs10232897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101363853 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCGCGGCTGCGGCTTTGTGTCGC[A/G]GAGCAGGTTCTCGGGTGGCCAAGGC
Phenotype
MIM: 608927
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COL26A1 PubMed Links

Gene Details

Gene
COL26A1
Gene Name
collagen type XXVI alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278563.2 Intron NP_001265492.1
NM_133457.4 Intron NP_597714.2
XM_017011743.1 Intron XP_016867232.1
XM_017011744.1 Intron XP_016867233.1
XM_017011745.1 Intron XP_016867234.1

View Full Product Details