Product Details

SNP ID
rs7303081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:133117610 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAAGCTTATATTAAACTTTATTA[A/T]GCTGCATATTAAACCTCACCATACT
Phenotype
MIM: 604082
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF140 PubMed Links

Gene Details

Gene
ZNF140
Gene Name
zinc finger protein 140
There are no transcripts associated with this gene.

Gene
ZNF891
Gene Name
zinc finger protein 891
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277291.1 4500 Intron NP_001264220.1
XM_017018666.1 4500 UTR 3 XP_016874155.1
XM_017018667.1 4500 UTR 3 XP_016874156.1
XM_017018668.1 4500 UTR 3 XP_016874157.1

View Full Product Details