Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001129896.2 | Intron | NP_001123368.1 | |||
NM_001174068.1 | Intron | NP_001167539.1 | |||
NM_080737.2 | Intron | NP_542775.2 | |||
XM_005262229.2 | Intron | XP_005262286.1 | |||
XM_011531068.2 | Intron | XP_011529370.1 | |||
XM_017029966.1 | Intron | XP_016885455.1 | |||
XM_017029967.1 | Intron | XP_016885456.1 | |||
XM_017029968.1 | Intron | XP_016885457.1 | |||
XM_017029969.1 | Intron | XP_016885458.1 | |||
XM_017029970.1 | Intron | XP_016885459.1 | |||
XM_017029971.1 | Intron | XP_016885460.1 |