Product Details

SNP ID
rs9645800
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:133118693 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAAAACTTTCTGTAGTTCCAATG[C/G]TGAAATGTTGGGATACTCTTTTTGC
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ZNF891 PubMed Links

Gene Details

Gene
ZNF891
Gene Name
zinc finger protein 891
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277291.1 3417 Intron NP_001264220.1
XM_017018666.1 3417 UTR 3 XP_016874155.1
XM_017018667.1 3417 UTR 3 XP_016874156.1
XM_017018668.1 3417 UTR 3 XP_016874157.1

View Full Product Details