Product Details
- SNP ID
-
rs9487887
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:112322108 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTCAAGCTCCCACACTGTGAAATA[C/T]TTGAACTCTCTAGGAAATATCCTGG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RFPL4B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76362291] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RFPL4B
- Gene Name
- ret finger protein like 4B
View Full Product Details