Product Details

SNP ID
rs10521774
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:132039131 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACATGCCTGGGTTCTGGGTCAAT[C/G]TGTATAACTAGACGTATCTTAATTT
Phenotype
MIM: 300547
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
STK26 PubMed Links

Gene Details

Gene
STK26
Gene Name
serine/threonine protein kinase 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042452.1 Intron NP_001035917.1
NM_001042453.1 Intron NP_001035918.1
NM_016542.3 Intron NP_057626.2
XM_011531349.1 Intron XP_011529651.1
XM_011531350.1 Intron XP_011529652.1

View Full Product Details