Product Details
- SNP ID
-
hCV30592448
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:35438233 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATTCCCTCAACTCCATTCATAACT[C/G]TCTTGCACCAAACACTTGATCTCCA
- Phenotype
-
MIM: 613535
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA0319L
PubMed Links
Gene Details
- Gene
- KIAA0319L
- Gene Name
- KIAA0319 like
There are no transcripts associated with this gene.
View Full Product Details