Product Details

SNP ID
rs13289988
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:99979689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTGGTTCTGGGGACTCAACCGT[C/G]TTCTTCAGTCTGGTCTTGCATCCTC
Phenotype
MIM: 609170 MIM: 604204
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ERP44 PubMed Links

Gene Details

Gene
ERP44
Gene Name
endoplasmic reticulum protein 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015051.2 4356 UTR 3 NP_055866.1
Gene
STX17
Gene Name
syntaxin 17
There are no transcripts associated with this gene.

View Full Product Details