Product Details

SNP ID
rs12322454
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:133119787 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAGAGGACAAGATAGATAGTTCTG[A/T]TCCCAGTGATATGTCTATTTTTGAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF891 PubMed Links

Gene Details

Gene
ZNF891
Gene Name
zinc finger protein 891
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277291.1 2323 Intron NP_001264220.1
XM_017018666.1 2323 UTR 3 XP_016874155.1
XM_017018667.1 2323 UTR 3 XP_016874156.1
XM_017018668.1 2323 UTR 3 XP_016874157.1

View Full Product Details