Product Details

SNP ID
rs12111900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:130346080 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGGATATGGCCCAGGGAAAGCAT[A/G]AGTGGGCTAGGCGGAGAAGAAGAGC
Phenotype
MIM: 609561
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPA5 PubMed Links

Gene Details

Gene
CPA5
Gene Name
carboxypeptidase A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127441.1 Intron NP_001120913.1
NM_001127442.1 Intron NP_001120914.1
NM_001318223.1 Intron NP_001305152.1
NM_080385.4 Intron NP_525124.3
XM_005250710.1 Intron XP_005250767.1
XM_005250712.1 Intron XP_005250769.1
XM_011516698.1 Intron XP_011515000.1
XM_011516701.1 Intron XP_011515003.1
XM_011516702.1 Intron XP_011515004.1
XM_011516703.1 Intron XP_011515005.1
XM_011516704.1 Intron XP_011515006.1
XM_011516705.1 Intron XP_011515007.1
Gene
LOC101928632
Gene Name
uncharacterized LOC101928632
There are no transcripts associated with this gene.

View Full Product Details