Product Details

SNP ID
rs11629184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:29578270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTAAGAAAACTCAGTGATTATTG[A/T]TTACCTGTAGCCAAGGGTGGCTCAA
Phenotype
MIM: 605435
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PRKD1 PubMed Links

Gene Details

Gene
PRKD1
Gene Name
protein kinase D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002742.2 Intron NP_002733.2
XM_005267859.1 Intron XP_005267916.1
XM_011536964.2 Intron XP_011535266.1
XM_011536965.2 Intron XP_011535267.1
XM_017021461.1 Intron XP_016876950.1
XM_017021462.1 Intron XP_016876951.1

View Full Product Details