Product Details
- SNP ID
-
rs7051133
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:73795932 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCTATGTACATATATCCTTTATAGG[A/G]AATTAAGGAGGTTATAGACACAACC
- Phenotype
-
MIM: 300181
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TSIX
PubMed Links
Gene Details
- Gene
- TSIX
- Gene Name
- TSIX transcript, XIST antisense RNA
There are no transcripts associated with this gene.
View Full Product Details