Product Details
- SNP ID
-
rs12898722
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:68059398 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTTAACTAAATATATTGATATGAC[A/T]GATCTTTTTTGTGCTTTAGAAAGCA
- Phenotype
-
MIM: 603566
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PIAS1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116918982] are located under a probe and SNP(s) [rs78364021] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PIAS1
- Gene Name
- protein inhibitor of activated STAT 1
View Full Product Details