Product Details
- SNP ID
-
rs17009013
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:84494270 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTAAAGTGAAGACTAGAATTACCA[A/T]TGGTTATTTTTAAAACGTGAAAAAT
- Phenotype
-
MIM: 602563
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NKX6-1
PubMed Links
Gene Details
- Gene
- NKX6-1
- Gene Name
- NK6 homeobox 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006168.2 |
|
Intron |
|
|
NP_006159.2 |
View Full Product Details