Product Details
- SNP ID
-
rs17328447
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:87630830 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGCTTTAGGTCATGTAATGATCA[C/T]GTGTGCCCTTATCAGAGACAGTTGA
- Phenotype
-
MIM: 171050
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ABCB1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs28381767] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABCB1
- Gene Name
- ATP binding cassette subfamily B member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000927.4 |
|
Intron |
|
|
NP_000918.2 |
- Gene
- RUNDC3B
- Gene Name
- RUN domain containing 3B
View Full Product Details