Product Details

SNP ID
rs17083519
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:26259970 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGTATATGGCAACACAGAGAAGG[A/G]AGTCCCCAACTGGTTAAGGTTTTTA
Phenotype
MIM: 603184
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDK8 PubMed Links

Gene Details

Gene
CDK8
Gene Name
cyclin dependent kinase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001260.2 Intron NP_001251.1
NM_001318368.1 Intron NP_001305297.1
XM_005266216.4 Intron XP_005266273.1
XM_011534864.1 Intron XP_011533166.1
XM_011534865.2 Intron XP_011533167.1
XM_017020317.1 Intron XP_016875806.1
XM_017020318.1 Intron XP_016875807.1

View Full Product Details