Product Details
- SNP ID
-
rs3825148
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:122716473 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCCAAACTTCCAGTCTGAACGCC[G/T]CACATAGTAGTCCATCACGAACGGC
- Phenotype
-
MIM: 606039
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HCAR3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11059549,rs1696352] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HCAR3
- Gene Name
- hydroxycarboxylic acid receptor 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006018.2 |
420 |
Silent Mutation |
|
|
NP_006009.2 |
View Full Product Details