Product Details

SNP ID
rs3008619
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:222626022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAGTGCTAGGATTACAGGCTGC[A/C]TTTCATATATATATATAAAAAACAC
Phenotype
MIM: 613455
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MIA3 PubMed Links

Gene Details

Gene
MIA3
Gene Name
MIA family member 3, ER export factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300867.1 Intron NP_001287796.1
NM_001324062.1 Intron NP_001310991.1
NM_001324063.1 Intron NP_001310992.1
NM_001324064.1 Intron NP_001310993.1
NM_001324065.1 Intron NP_001310994.1
NM_198551.3 Intron NP_940953.2
XM_006711304.3 Intron XP_006711367.2
XM_011509513.2 Intron XP_011507815.2
XM_017001243.1 Intron XP_016856732.1

View Full Product Details