Product Details

SNP ID
rs34412722
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:172768130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGCCATAATTTATTCCATTGTA[C/T]AAAAAAGTCATCCTTATGTAACAAA
Phenotype
MIM: 600714
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DUSP1 PubMed Links

Gene Details

Gene
DUSP1
Gene Name
dual specificity phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004417.3 1984 UTR 3 NP_004408.1
Gene
LOC101928093
Gene Name
uncharacterized LOC101928093
There are no transcripts associated with this gene.

View Full Product Details