Product Details

SNP ID
rs58747475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:93244935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTAAATAATACAGAAGTTAACTA[C/T]AGAGGAAGCCTCACTGGATGGTGTT
Phenotype
MIM: 602190
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EPHA7 PubMed Links

Gene Details

Gene
EPHA7
Gene Name
EPH receptor A7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288629.1 Intron NP_001275558.1
NM_001288630.1 Intron NP_001275559.1
NM_004440.3 Intron NP_004431.1
XM_005248669.2 Intron XP_005248726.1
XM_005248671.2 Intron XP_005248728.1
XM_017010365.1 Intron XP_016865854.1
XM_017010366.1 Intron XP_016865855.1

View Full Product Details