Product Details

SNP ID
rs61017022
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88876666 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGGTGGGCAGAGCCGCCGCGCC[C/T]GCGGCATCTGCTCTGCTGTCTAAAG
Phenotype
MIM: 603870
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CBFA2T3 PubMed Links

Gene Details

Gene
CBFA2T3
Gene Name
CBFA2/RUNX1 translocation partner 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005187.5 2227 UTR 3 NP_005178.4
NM_175931.2 2227 UTR 3 NP_787127.1
XM_005256323.4 2227 UTR 3 XP_005256380.1
XM_011523419.2 2227 Intron XP_011521721.1
XM_017023811.1 2227 Intron XP_016879300.1
Gene
LOC101927793
Gene Name
uncharacterized LOC101927793
There are no transcripts associated with this gene.

Gene
PABPN1L
Gene Name
poly(A) binding protein nuclear 1 like (cytoplasmic)
There are no transcripts associated with this gene.

View Full Product Details