Product Details

SNP ID
rs56361301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:134650284 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAGTGATCTCCCAGTGAGAACAC[A/G]TGAGCATCGTAACCGTCTTCCATTG
Phenotype
MIM: 120215
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COL5A1 PubMed Links

Gene Details

Gene
COL5A1
Gene Name
collagen type V alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000093.4 Intron NP_000084.3
NM_001278074.1 Intron NP_001265003.1
XM_017014266.1 Intron XP_016869755.1
Gene
COL5A1-AS1
Gene Name
COL5A1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details