Product Details

SNP ID
rs73156835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:139564680 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGCCTTGGGGGAGACACAACCT[C/T]CAACTAAGAGGCATTCACTGGGGTT
Phenotype
MIM: 606868
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HIPK2 PubMed Links

Gene Details

Gene
HIPK2
Gene Name
homeodomain interacting protein kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113239.2 12610 UTR 3 NP_001106710.1
NM_022740.4 12610 UTR 3 NP_073577.3
XM_006715935.2 12610 UTR 3 XP_006715998.1
XM_011516077.2 12610 UTR 3 XP_011514379.1
XM_011516078.2 12610 UTR 3 XP_011514380.1
XM_011516079.2 12610 UTR 3 XP_011514381.1
XM_011516080.2 12610 UTR 3 XP_011514382.1
XM_011516081.2 12610 UTR 3 XP_011514383.1
XM_017012069.1 12610 UTR 3 XP_016867558.1

View Full Product Details