Product Details

SNP ID
rs72943263
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:56608334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAAACAATGAAATACTAGCTTA[C/T]TTTTAAAGAATAACCCTAAAAGACA
Phenotype
MIM: 603049
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TXNL1 PubMed Links

Gene Details

Gene
TXNL1
Gene Name
thioredoxin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004786.2 Intron NP_004777.1
XM_006722580.3 Intron XP_006722643.1
XM_011526263.2 Intron XP_011524565.1
XM_011526264.2 Intron XP_011524566.1
XM_017026093.1 Intron XP_016881582.1
XM_017026094.1 Intron XP_016881583.1

View Full Product Details