Product Details

SNP ID
rs75670763
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:168082316 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGCAGGCTAGCCAGGTGGTCCTT[A/G]GTCACCAAGGCCGTGGACTACACGA
Phenotype
MIM: 610494 MIM: 612250
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCAF6 PubMed Links

Gene Details

Gene
DCAF6
Gene Name
DDB1 and CUL4 associated factor 6
There are no transcripts associated with this gene.

Gene
GPR161
Gene Name
G protein-coupled receptor 161
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267609.1 5372 UTR 3 NP_001254538.1
NM_001267610.1 5372 UTR 3 NP_001254539.1
NM_001267611.1 5372 UTR 3 NP_001254540.1
NM_001267612.1 5372 UTR 3 NP_001254541.1
NM_001267613.1 5372 UTR 3 NP_001254542.1
NM_001267614.1 5372 UTR 3 NP_001254543.1
NM_153832.2 5372 UTR 3 NP_722561.1
XM_005245055.2 5372 UTR 3 XP_005245112.1
XM_005245056.2 5372 UTR 3 XP_005245113.1
XM_005245057.4 5372 UTR 3 XP_005245114.1
XM_006711251.1 5372 UTR 3 XP_006711314.1
XM_006711253.2 5372 UTR 3 XP_006711316.2
XM_011509370.1 5372 UTR 3 XP_011507672.1
XM_011509371.2 5372 UTR 3 XP_011507673.1
XM_011509372.1 5372 UTR 3 XP_011507674.1
XM_011509373.2 5372 UTR 3 XP_011507675.1
XM_011509375.2 5372 UTR 3 XP_011507677.1
XM_011509376.2 5372 UTR 3 XP_011507678.1
XM_011509377.1 5372 UTR 3 XP_011507679.1
XM_011509378.1 5372 UTR 3 XP_011507680.1
XM_017000858.1 5372 UTR 3 XP_016856347.1
XM_017000859.1 5372 UTR 3 XP_016856348.1
XM_017000860.1 5372 UTR 3 XP_016856349.1
XM_017000861.1 5372 UTR 3 XP_016856350.1
XM_017000862.1 5372 UTR 3 XP_016856351.1

View Full Product Details