Product Details

SNP ID
rs16931769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:27253814 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCATATTAGGGGGCCACAGGAACC[A/G]GAATTGTTGCCGTGATGCTGGACAG
Phenotype
MIM: 615836
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STK38L PubMed Links

Gene Details

Gene
STK38L
Gene Name
serine/threonine kinase 38 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015000.3 Intron NP_055815.1
XM_005253342.3 Intron XP_005253399.1
XM_006719058.3 Intron XP_006719121.1
XM_006719059.3 Intron XP_006719122.1
XM_011520613.2 Intron XP_011518915.1
XM_017019036.1 Intron XP_016874525.1

View Full Product Details