Product Details

SNP ID
rs2290108
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:113713201 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAATTGAACAGTGGTGCTTCCTTCT[A/G]AAGAAGTTGTTCATTGATTTTTGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR4782 PubMed Links

Gene Details

Gene
MIR4782
Gene Name
microRNA 4782
There are no transcripts associated with this gene.

Gene
SLC35F5
Gene Name
solute carrier family 35 member F5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005263799.3 4056 UTR 3 XP_005263856.1
XM_011511921.2 4056 UTR 3 XP_011510223.1
XM_011511922.2 4056 UTR 3 XP_011510224.1
XM_011511923.2 4056 UTR 3 XP_011510225.1
XM_017005027.1 4056 Intron XP_016860516.1
XM_017005028.1 4056 UTR 3 XP_016860517.1

View Full Product Details