Product Details

SNP ID
rs34832868
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:113713023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTATTTCTTGAGAAGATGAACC[C/T]TTAACTATGAAGGTGCAGAAAGGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4782 PubMed Links

Gene Details

Gene
MIR4782
Gene Name
microRNA 4782
There are no transcripts associated with this gene.

Gene
SLC35F5
Gene Name
solute carrier family 35 member F5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005263799.3 4234 UTR 3 XP_005263856.1
XM_011511921.2 4234 UTR 3 XP_011510223.1
XM_011511922.2 4234 UTR 3 XP_011510224.1
XM_011511923.2 4234 UTR 3 XP_011510225.1
XM_017005027.1 4234 Intron XP_016860516.1
XM_017005028.1 4234 UTR 3 XP_016860517.1

View Full Product Details